siRNA Details
| Gene Symbol: Olfr586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147111.1 | NP_667322.1 | 1 | 718 | ||
| GenBank mRNA | AF121985.1 | - | 1 | 320 | ||
| BC104726.1 | - | 1 | 717 | 40043611 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr586| Gene Symbol: Olfr586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147111.1 | NP_667322.1 | 1 | 718 | ||
| GenBank mRNA | AF121985.1 | - | 1 | 320 | ||
| BC104726.1 | - | 1 | 717 | 40043611 | ||