siRNA Details
| Gene Symbol: Olfr1353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147042.2 | NP_667253.1 | 3 | 598 | ||
| GenBank mRNA | BC119112.1 | - | 1 | 347 | 8733861 | |
| BC120824.1 | - | 1 | 347 | 40129747 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1353| Gene Symbol: Olfr1353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147042.2 | NP_667253.1 | 3 | 598 | ||
| GenBank mRNA | BC119112.1 | - | 1 | 347 | 8733861 | |
| BC120824.1 | - | 1 | 347 | 40129747 | ||