siRNA Details
| Gene Symbol: Olfr630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147098.2 | NP_667309.1 | 2 | 1029 | ||
| GenBank mRNA | BC116998.1 | - | 1 | 987 | 40126317 | |
| BC117000.1 | - | 1 | 987 | 40126319 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr630| Gene Symbol: Olfr630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147098.2 | NP_667309.1 | 2 | 1029 | ||
| GenBank mRNA | BC116998.1 | - | 1 | 987 | 40126317 | |
| BC117000.1 | - | 1 | 987 | 40126319 | ||