siRNA Details
| Gene Symbol: Olfr635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147118.2 | NP_667329.2 | 1 | 281 | ||
| GenBank mRNA | BC152334.1 | - | 1 | 285 | ||
| BC152355.1 | - | 1 | 285 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr635| Gene Symbol: Olfr635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147118.2 | NP_667329.2 | 1 | 281 | ||
| GenBank mRNA | BC152334.1 | - | 1 | 285 | ||
| BC152355.1 | - | 1 | 285 | |||