siRNA Details
Gene Symbol: Olfr632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147119.1 | NP_667330.1 | 1 | 935 | ||
GenBank mRNA | BC106823.2 | - | 1 | 935 | 40043471 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147119.1 | NP_667330.1 | 1 | 935 | ||
GenBank mRNA | BC106823.2 | - | 1 | 935 | 40043471 |