siRNA Details
| Gene Symbol: Olfr582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147053.1 | NP_667264.1 | 1 | 945 | ||
| GenBank mRNA | BC107338.1 | - | 1 | 945 | ||
| BC107339.1 | - | 1 | 945 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr582| Gene Symbol: Olfr582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147053.1 | NP_667264.1 | 1 | 945 | ||
| GenBank mRNA | BC107338.1 | - | 1 | 945 | ||
| BC107339.1 | - | 1 | 945 | |||