siRNA Details
| Gene Symbol: Olfr649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147055.1 | NP_667266.1 | 1 | 523 | ||
| GenBank mRNA | BC104080.2 | - | 1 | 516 | ||
| BC104081.2 | - | 1 | 516 | |||
| BC119409.2 | - | 1 | 516 | |||
| BC119410.2 | - | 1 | 516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr649| Gene Symbol: Olfr649 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147055.1 | NP_667266.1 | 1 | 523 | ||
| GenBank mRNA | BC104080.2 | - | 1 | 516 | ||
| BC104081.2 | - | 1 | 516 | |||
| BC119409.2 | - | 1 | 516 | |||
| BC119410.2 | - | 1 | 516 | |||