siRNA Details
| Gene Symbol: Olfr449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147064.1 | NP_667275.1 | 1 | 890 | ||
| GenBank mRNA | BC138166.1 | - | 1 | 921 | 8861185 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr449| Gene Symbol: Olfr449 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147064.1 | NP_667275.1 | 1 | 890 | ||
| GenBank mRNA | BC138166.1 | - | 1 | 921 | 8861185 | |