siRNA Details
| Gene Symbol: Olfr559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147112.1 | NP_667323.1 | 1 | 945 | ||
| GenBank mRNA | BC119185.1 | - | 1 | 967 | 8733934 | |
| BC120640.1 | - | 1 | 967 | 40129563 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr559| Gene Symbol: Olfr559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147112.1 | NP_667323.1 | 1 | 945 | ||
| GenBank mRNA | BC119185.1 | - | 1 | 967 | 8733934 | |
| BC120640.1 | - | 1 | 967 | 40129563 | ||