siRNA Details
| Gene Symbol: Olfr638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147120.1 | NP_667331.1 | 1 | 281 | ||
| GenBank mRNA | BC128011.1 | - | 1 | 280 | 40136021 | |
| BC152352.1 | - | 1 | 285 | 40136020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr638| Gene Symbol: Olfr638 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147120.1 | NP_667331.1 | 1 | 281 | ||
| GenBank mRNA | BC128011.1 | - | 1 | 280 | 40136021 | |
| BC152352.1 | - | 1 | 285 | 40136020 | ||