siRNA Details
Gene Symbol: Olfr644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147121.1 | NP_667332.1 | 1 | 216 | ||
GenBank mRNA | BC099886.1 | - | 1 | 208 | ||
BC099887.1 | - | 1 | 208 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr644 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147121.1 | NP_667332.1 | 1 | 216 | ||
GenBank mRNA | BC099886.1 | - | 1 | 208 | ||
BC099887.1 | - | 1 | 208 |