siRNA Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148673.3 | NP_683514.2 | 3 | 261 | ||
| NM_198936.1 | NP_945174.1 | 2 | 182 | |||
| GenBank mRNA | AK029117.1 | - | 2 | 152 | ||
| AK046262.1 | - | 2 | 153 | |||
| AK047597.1 | - | 3 | 249 | |||
| AK049178.1 | - | 2 | 151 | |||
| BC025870.1 | - | 3 | 240 | |||
| BC060954.1 | - | 2 | 150 | |||
| BC082780.1 | - | 3 | 241 | |||
| BC106099.1 | - | 2 | 163 | |||
| BY243619.1 | - | 2 | 185 | |||

Slu7
Genomic Map