siRNA Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029838.2 | NP_001025009.1 | 6 | 641 | ||
| NM_148950.3 | NP_683752.2 | 5 | 536 | |||
| GenBank mRNA | AB086817.1 | - | 3 | 362 | ||
| AF487460.1 | - | 4 | 442 | |||
| AK082952.1 | - | 6 | 586 | |||
| AK084178.1 | - | 6 | 584 | |||
| AK144270.1 | - | 6 | 578 | |||
| BC050865.1 | - | 5 | 470 | 6332968 | ||

Pknox2
Genomic Map