siRNA Details
| Gene Symbol: Asb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148953.2 | NP_683755.1 | 2 | 512 | ||
| GenBank mRNA | AF403044.1 | - | 2 | 512 | ||
| BC127245.1 | - | 2 | 508 | |||
| BC127246.1 | - | 2 | 511 | |||
| BC152348.1 | - | 2 | 512 | |||
| BC152349.1 | - | 2 | 512 | 40104909 | ||

Asb16
Genomic Map