siRNA Details
| Gene Symbol: Pop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026340.3 | NP_080616.1 | 3 | 279 | ||
| NM_152894.2 | NP_690854.1 | 3 | 279 | |||
| GenBank mRNA | AK016547.1 | - | 3 | 266 | ||
| AK018297.1 | - | 3 | 248 | |||
| BC034742.1 | - | 3 | 258 | 4935194 | ||

Pop1
Genomic Map