siRNA Details
| Gene Symbol: Zfp472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153063.3 | NP_694703.2 | 2 | 206 | ||
| GenBank mRNA | AK142395.1 | - | 2 | 197 | ||
| AY195875.1 | - | 2 | 141 | |||
| BB652931.1 | - | 2 | 207 | |||
| BC023090.1 | - | 2 | 141 | |||
| BC027407.1 | - | 2 | 196 | 4945483 | ||

Zfp472
Genomic Map