siRNA Details
| Gene Symbol: Olfr644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147121.1 | NP_667332.1 | 1 | 524 | ||
| GenBank mRNA | BC099886.1 | - | 1 | 516 | ||
| BC099887.1 | - | 1 | 516 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr644| Gene Symbol: Olfr644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147121.1 | NP_667332.1 | 1 | 524 | ||
| GenBank mRNA | BC099886.1 | - | 1 | 516 | ||
| BC099887.1 | - | 1 | 516 | |||