siRNA Details
| Gene Symbol: Vwa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147776.4 | NP_680085.3 | 4 | 1614 | ||
| GenBank mRNA | AK030019.1 | - | 4 | 1568 | ||
| AK039219.1 | - | 2 | 738 | |||
| AK077240.1 | - | 4 | 1585 | |||
| AY030094.1 | - | 4 | 1584 | |||
| BC020136.1 | - | 2 | 921 | |||
| BC026919.1 | - | 4 | 1572 | 3982749 | ||
| BC036166.1 | - | 4 | 1582 | 4985003 | ||

Vwa1
Genomic Map