siRNA Details
| Gene Symbol: Zfp263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148924.3 | NP_683726.2 | 2 | 832 | ||
| GenBank mRNA | AF499776.1 | - | 2 | 800 | ||
| AK004765.2 | - | 2 | 829 | |||
| AK159455.1 | - | 2 | 820 | |||
| AK182706.1 | - | 2 | 186 | |||
| BC011505.1 | - | 3 | 1358 | |||
| BC056222.1 | - | 2 | 653 | 30009326 | ||
| BC060162.1 | - | 1 | 1765 | |||

Zfp263
Genomic Map