siRNA Details
| Gene Symbol: Gm9125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163730.2 | NP_001157202.1 | 2 | 338 | ||
| GenBank mRNA | AK033449.1 | - | 2 | 339 | ||
| Gene Symbol: Tdpoz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148949.2 | NP_683751.2 | 2 | 311 | ||
| GenBank mRNA | AF290198.1 | - | 2 | 296 | ||
| BC125302.1 | - | 1 | 173 | 40129817 | ||
| BC125304.1 | - | 1 | 173 | 40129819 | ||

Gm9125
Genomic Map