siRNA Details
| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152809.2 | NP_690022.2 | 6 | 1312 | ||
| GenBank mRNA | AK218008.1 | - | 3 | 362 | ||
| AK218677.1 | - | 3 | 338 | |||
| BC033601.1 | - | 3 | 327 | |||
| BQ443928.1 | - | 3 | 346 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csnk1g3| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152809.2 | NP_690022.2 | 6 | 1312 | ||
| GenBank mRNA | AK218008.1 | - | 3 | 362 | ||
| AK218677.1 | - | 3 | 338 | |||
| BC033601.1 | - | 3 | 327 | |||
| BQ443928.1 | - | 3 | 346 | |||