siRNA Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290425.1 | NP_001277354.1 | 5, 6 | 544 | ||
| NM_152825.2 | NP_690038.1 | 5, 6 | 544 | |||
| GenBank mRNA | AK019756.1 | - | 6, 7 | 659 | ||
| AK088073.1 | - | 5, 6 | 527 | |||
| AK162825.1 | - | 5, 6 | 545 | |||
| BC027768.1 | - | 5, 6 | 527 | 5354502 | ||

Usp45
Genomic Map