siRNA Details
| Gene Symbol: Gzmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB049454.2 | - | 4 | 362 | ||
| AB100426.1 | - | 4 | 362 | |||
| BC120809.1 | - | 3 | 283 | 40129732 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gzmn| Gene Symbol: Gzmn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB049454.2 | - | 4 | 362 | ||
| AB100426.1 | - | 4 | 362 | |||
| BC120809.1 | - | 3 | 283 | 40129732 | ||