siRNA Details
| Gene Symbol: Nrbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147201.2 | NP_671734.1 | 10, 11 | 1014 | ||
| GenBank mRNA | AF302138.1 | - | 10, 11 | 1013 | ||
| AK046142.1 | - | 11, 12 | 1032 | |||
| AK077440.1 | - | 10, 11 | 1050 | |||
| AK170501.1 | - | 10, 11 | 1030 | |||
| AK188331.1 | - | 2, 3 | 155 | |||
| AK207083.1 | - | 2, 3 | 155 | |||
| BC004756.1 | - | 10, 11 | 984 | 3154089 | ||
| BC018463.1 | - | 10, 11 | 988 | 3598438 | ||

Nrbp1
Genomic Map