siRNA Details
| Gene Symbol: Rdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147222.2 | NP_671755.2 | 4 | 1397 | ||
| GenBank mRNA | AY046408.1 | - | 4 | 1394 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh19| Gene Symbol: Rdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147222.2 | NP_671755.2 | 4 | 1397 | ||
| GenBank mRNA | AY046408.1 | - | 4 | 1394 | ||