siRNA Details
| Gene Symbol: Myo3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148413.3 | NP_680779.3 | 267 | |||
| GenBank mRNA | AK018511.1 | - | 266 | |||
| AY101368.1 | - | 110 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo3a| Gene Symbol: Myo3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148413.3 | NP_680779.3 | 267 | |||
| GenBank mRNA | AK018511.1 | - | 266 | |||
| AY101368.1 | - | 110 | ||||