siRNA Details
| Gene Symbol: Slu7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148673.3 | NP_683514.2 | 17 | 2457 | ||
| NM_198936.1 | NP_945174.1 | 16 | 2378 | |||
| GenBank mRNA | AK029117.1 | - | 16 | 2348 | ||
| AK046262.1 | - | 16 | 2349 | |||
| BC013810.1 | - | 14 | 1977 | |||
| BC023057.1 | - | 6 | 1232 | |||

Slu7
Genomic Map