siRNA Details
| Gene Symbol: Rps6ka3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_148945.2 | NP_683747.1 | 9 | 994 | ||
| GenBank mRNA | AK079102.1 | - | 1087 | |||
| AK154778.1 | - | 9 | 907 | |||
| AY083469.1 | - | 9 | 656 | |||
| BC150156.1 | - | 9 | 659 | |||
| BC150478.1 | - | 9 | 659 | |||

Rps6ka3
Genomic Map