siRNA Details
| Gene Symbol: Csnk1g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152809.2 | NP_690022.2 | 13 | 3718 | ||
| GenBank mRNA | AK032675.1 | - | 7 | 2273 | ||
| AK034842.1 | - | 3 | 1899 | |||
| BC025921.1 | - | 1 | 901 | |||
| BC033601.1 | - | 10 | 2733 | |||

Csnk1g3
Genomic Map