siRNA Details
| Gene Symbol: Purg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098233.1 | NP_001091703.1 | 1 | 618 | ||
| NM_152821.2 | NP_690034.1 | 1 | 618 | |||
| GenBank mRNA | AF479672.1 | - | 1 | 618 | ||
| AF479673.1 | - | 1 | 618 | |||
| AK015630.1 | - | 2 | 630 | |||
| BC116406.2 | - | 1 | 618 | 40102393 | ||
| CD772246.1 | - | 1 | 183 | |||

Purg
Genomic Map