siRNA Details
| Gene Symbol: Usp45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290425.1 | NP_001277354.1 | 6 | 640 | ||
| NM_152825.2 | NP_690038.1 | 6 | 640 | |||
| GenBank mRNA | AK019756.1 | - | 7 | 755 | ||
| AK088073.1 | - | 6 | 623 | |||
| AK162825.1 | - | 6 | 641 | |||
| BC027768.1 | - | 6 | 623 | 5354502 | ||

Usp45
Genomic Map