siRNA Details
| Gene Symbol: Jchain | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152839.3 | NP_690052.2 | 4 | 1055 | ||
| GenBank mRNA | AK078865.1 | - | 4 | 942 | ||
| AK160344.1 | - | 4 | 1008 | |||
| BC006026.1 | - | 4 | 962 | 3491646 | ||
| J00544.1 | - | 4 | 937 | |||

Jchain
Genomic Map