siRNA Details
| Gene Symbol: Sec63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153055.3 | NP_694695.3 | 21 | 2726 | ||
| GenBank mRNA | AF397910.1 | - | 7 | 1006 | ||
| AK036629.1 | - | 21 | 2417 | |||
| AY024346.1 | - | 21 | 2523 | |||
| AY238936.1 | - | 21 | 2496 | |||
| BC019366.1 | - | 21 | 2490 | 5066908 | ||
| BC029774.1 | - | 13 | 1727 | |||
| BC031846.1 | - | 21 | 2490 | 5066822 | ||
| BC059816.1 | - | 21 | 2725 | |||
| BC080810.1 | - | 20 | 4027 | |||

Sec63
Genomic Map