siRNA Details
| Gene Symbol: Ehbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252515.1 | NP_001239444.1 | 2 | 480 | ||
| NM_153078.4 | NP_694718.4 | 2 | 480 | |||
| GenBank mRNA | AF424697.1 | - | 1 | 330 | ||
| AK173055.1 | - | 2 | 481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehbp1| Gene Symbol: Ehbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252515.1 | NP_001239444.1 | 2 | 480 | ||
| NM_153078.4 | NP_694718.4 | 2 | 480 | |||
| GenBank mRNA | AF424697.1 | - | 1 | 330 | ||
| AK173055.1 | - | 2 | 481 | |||