siRNA Details
Gene Symbol: Mrgpra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153101.3 | NP_694741.2 | 2 | 462 | ||
GenBank mRNA | AK156116.1 | - | 2 | 468 | ||
AY042192.1 | - | 436 |
Gene Symbol: Mrgpra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172588.1 | NP_001166059.1 | 437 | |||
GenBank mRNA | BC064040.1 | - | 437 | 1428847 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
