siRNA Details
| Gene Symbol: Zfp352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153102.3 | NP_694742.2 | 2 | 359 | ||
| GenBank mRNA | AF290196.1 | - | 2 | 359 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp352| Gene Symbol: Zfp352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153102.3 | NP_694742.2 | 2 | 359 | ||
| GenBank mRNA | AF290196.1 | - | 2 | 359 | ||