siRNA Details
| Gene Symbol: Traf3ip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153137.4 | NP_694777.3 | 3 | 627 | ||
| GenBank mRNA | AK031400.1 | - | 3 | 567 | ||
| AK036608.1 | - | 3 | 396 | |||
| AK041994.1 | - | 3 | 628 | |||
| AK080080.1 | - | 3 | 597 | |||
| AK161211.1 | - | 1 | 14 | |||
| AY383616.1 | - | 1 | 210 | |||
| BC036561.1 | - | 3 | 435 | |||
| BC137605.1 | - | 1 | 238 | 8860624 | ||
| BC171933.1 | - | 1 | 238 | 9054217 | ||

Traf3ip3
Genomic Map