siRNA Details
| Gene Symbol: Kctd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153143.4 | NP_694783.1 | 1 | 957 | ||
| GenBank mRNA | AF465352.1 | - | 1 | 924 | ||
| AK039558.1 | - | 1379 | ||||
| AK161602.1 | - | 1 | 952 | |||
| AK170258.1 | - | 1 | 851 | |||
| BC052502.1 | - | 1 | 205 | 5119264 | ||
| BC052734.1 | - | 1 | 908 | 6816736 | ||
| BC063052.1 | - | 1 | 908 | |||

Kctd11
Genomic Map