siRNA Details
| Gene Symbol: Zfp740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289690.1 | NP_001276619.1 | 6 | 1092 | ||
| NM_001289691.1 | NP_001276620.1 | 8 | 1291 | |||
| NM_001289694.1 | NP_001276623.1 | 7 | 1000 | |||
| NM_001289695.1 | NP_001276624.1 | 7 | 966 | |||
| GenBank mRNA | AK144707.1 | - | 8 | 1288 | ||
| AK170207.1 | - | 7 | 988 | |||
| BC030410.1 | - | 3 | 797 | 5367159 | ||
| BC066011.1 | - | 7 | 860 | 6418561 | ||
| CA319591.1 | - | 6 | 630 | |||

Zfp740
Genomic Map