siRNA Details
| Gene Symbol: Fbxo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153195.2 | NP_694875.2 | 2 | 261 | ||
| GenBank mRNA | AK147797.1 | - | 2 | 213 | ||
| AK147907.1 | - | 2 | 180 | |||
| AK150626.1 | - | 2 | 219 | |||
| AK151025.1 | - | 2 | 219 | |||
| AK152513.1 | - | 2 | 219 | |||
| AK208289.1 | - | 1 | 58 | |||
| AK212373.1 | - | 1 | 58 | |||
| AK219779.1 | - | 1 | 57 | |||
| BC032153.1 | - | 2 | 203 | 5054087 | ||
| CD563204.1 | - | 2 | 261 | |||

Fbxo7
Genomic Map