siRNA Details
| Gene Symbol: Thtpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153083.5 | NP_694723.1 | 1 | 904 | ||
| GenBank mRNA | AF432864.2 | - | 1 | 735 | ||
| AK085173.1 | - | 1 | 599 | |||
| AK132479.1 | - | 1 | 596 | |||
| BC025562.1 | - | 1 | 888 | 5322971 | ||

Thtpa
Genomic Map