siRNA Details
| Gene Symbol: Sec16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153125.2 | NP_694765.2 | 16 | 5616 | ||
| GenBank mRNA | AK122242.2 | - | 9 | 1053 | ||
| BC034649.1 | - | 6 | 674 | |||
| BC042603.1 | - | 10 | 1132 | |||
| BC059898.1 | - | 7 | 1219 | |||

Sec16a
Genomic Map