siRNA Details
| Gene Symbol: Ggnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB064543.1 | - | 6 | 744 | ||
| BC145605.1 | - | 5 | 652 | 9054171 | ||
| BC150801.1 | - | 5 | 652 | 9087712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggnbp2| Gene Symbol: Ggnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB064543.1 | - | 6 | 744 | ||
| BC145605.1 | - | 5 | 652 | 9054171 | ||
| BC150801.1 | - | 5 | 652 | 9087712 | ||