siRNA Details
| Gene Symbol: 2410015M20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153152.3 | NP_694792.1 | 4 | 417 | ||
| GenBank mRNA | AK075942.1 | - | 4 | 348 | ||
| AK075961.1 | - | 4 | 418 | |||
| AY091637.1 | - | 4 | 360 | |||
| BC049674.1 | - | 238 | ||||
| BC056164.1 | - | 4 | 323 | 5713062 | ||

2410015M20Rik
Genomic Map