siRNA Details
| Gene Symbol: E130308A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015681.2 | NP_001015681.1 | 2 | 525 | ||
| NM_153158.5 | NP_694798.1 | 2 | 525 | |||
| GenBank mRNA | AK047478.1 | - | 2 | 526 | ||
| AK047601.1 | - | 2 | 494 | |||
| AK081604.1 | - | 2 | 409 | |||
| AK087509.1 | - | 2 | 522 | |||
| BC017649.1 | - | 2 | 292 | 4924105 | ||
| BC037070.1 | - | 2 | 343 | 5004205 | ||

E130308A19Rik
Genomic Map