siRNA Details
| Gene Symbol: BC037034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153161.3 | NP_694801.2 | 11 | 1902 | ||
| GenBank mRNA | AK046882.1 | - | 7 | 2925 | ||
| AK138960.1 | - | 11 | 1846 | |||
| AK213494.1 | - | 2 | 168 | |||
| AK216902.1 | - | 2 | 221 | |||
| BC037034.1 | - | 10 | 1305 | 4505093 | ||
| BC040828.1 | - | 1 | 1875 | |||
| BC044860.1 | - | 1 | 1887 | |||

BC037034
Genomic Map