siRNA Details
| Gene Symbol: Slc16a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153081.3 | NP_694721.2 | 5 | 1507 | ||
| GenBank mRNA | AB041591.1 | - | 4 | 1385 | ||
| BC130255.1 | - | 4 | 1010 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc16a11| Gene Symbol: Slc16a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153081.3 | NP_694721.2 | 5 | 1507 | ||
| GenBank mRNA | AB041591.1 | - | 4 | 1385 | ||
| BC130255.1 | - | 4 | 1010 | |||