siRNA Details
| Gene Symbol: Ppp1r16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159662.1 | NP_001153134.1 | 5, 6 | 793 | ||
| NM_153089.4 | NP_694729.1 | 5, 6 | 771 | |||
| GenBank mRNA | AF423761.1 | - | 5, 6 | 790 | ||
| AK037627.1 | - | 1088 | ||||
| AK042123.1 | - | 5, 6 | 768 | |||
| AK044453.1 | - | 5, 6 | 809 | |||
| BC054764.1 | - | 5, 6 | 792 | 6414610 | ||
| BC057542.1 | - | 5, 6 | 792 | 6414610 | ||

Ppp1r16b
Genomic Map