siRNA Details
| Gene Symbol: Fnbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114665.2 | NP_001108137.1 | 16 | 2611 | ||
| NM_153118.3 | NP_694758.2 | 14 | 2437 | |||
| GenBank mRNA | BC034510.1 | - | 9 | 1885 | 4481144 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnbp1l| Gene Symbol: Fnbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114665.2 | NP_001108137.1 | 16 | 2611 | ||
| NM_153118.3 | NP_694758.2 | 14 | 2437 | |||
| GenBank mRNA | BC034510.1 | - | 9 | 1885 | 4481144 | |