siRNA Details
| Gene Symbol: Oplah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153122.2 | NP_694762.1 | 22 | 3136 | ||
| GenBank mRNA | AK164524.1 | - | 6 | 760 | ||
| BC025120.1 | - | 9 | 1234 | 4020691 | ||
| BC034522.1 | - | 22 | 3136 | 4236444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oplah| Gene Symbol: Oplah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153122.2 | NP_694762.1 | 22 | 3136 | ||
| GenBank mRNA | AK164524.1 | - | 6 | 760 | ||
| BC025120.1 | - | 9 | 1234 | 4020691 | ||
| BC034522.1 | - | 22 | 3136 | 4236444 | ||